5-Formylfuran-2-boronic acid, pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carbaldehyde
A693690-1G
A693690-250MG
A693690-25G
A693690-5G
76831-574EA
12
USD
76831-574
77886-621
77886-622
77886-623
5-Formylfuran-2-boronic acid, pinacol ester 98%
5-Formylfuran-2-boronic acid, pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₅BO₄ |
MDL Number:
MFCD16295105 CAS Number: 273731-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...