6-Benzyltetrahydro-1H-pyrrolo[3,4-b]pyridine-5,7(6H,7aH)-dione 97%

Supplier: Ambeed
Warning

A692080-100MG A692080-250MG
76831-566EA 47.75 USD
76831-566 76831-568
6-Benzyltetrahydro-1H-pyrrolo[3,4-b]pyridine-5,7(6H,7aH)-dione 97%
6-Benzyltetrahydro-1H-pyrrolo[3,4-b]pyridine-5,7(6H,7aH)-dione

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₂
MW: 244.29 g/mol
MDL Number: MFCD06795495
CAS Number: 128740-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR