5-Bromo-7-fluoroisoindolin-1-one 97%

Supplier: Ambeed
Warning

A691391-100MG A691391-250MG A691391-1G A691391-5G
76831-556EA 39 USD
76831-556 76831-558 77308-130 77886-616
5-Bromo-7-fluoroisoindolin-1-one 97%
5-Bromo-7-fluoroisoindolin-1-one

Precaución: for Research Use Only

Formula: C₈H₅BrFNO
MW: 230.03 g/mol
MDL Number: MFCD12755860
CAS Number: 957346-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR