1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole

A689163-1G A689163-250MG A689163-5G
76831-548EA 51.99 USD
76831-548 76831-550 76831-552
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole 98%
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂
MDL Number: MFCD09817558
CAS Number: 62546-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR