5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene 98%
Supplier: Ambeed
|
Warning
|
A686319-100MG
A686319-1G
A686319-250MG
A686319-5G
76831-530EA
10
USD
76831-530
76831-532
76831-534
76831-536
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene 98%
5,5'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,2'-bithiophene
Precaución: for Research Use Only
Formula:
C₂₀H₂₈B₂O₄S₂ |
MDL Number:
MFCD06798112 CAS Number: 239075-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...