2,2,2-Trifluoro-1-(2-(methylamino)phenyl)ethanone 98%

Supplier: Ambeed
Warning

A683231-100MG A683231-1G A683231-250MG
76831-508EA 30.77 USD
76831-508 76831-510 76831-512
2,2,2-Trifluoro-1-(2-(methylamino)phenyl)ethanone 98%
2,2,2-Trifluoro-1-(2-(methylamino)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₃NO
MW: 203.16 g/mol
MDL Number: MFCD13172937
CAS Number: 202270-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR