2-Fluoro-5-bromobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-fluorobenzyl Bromide

A674896-25G A674896-5G
76831-482EA 109.28 USD
76831-482 77684-580
2-Fluoro-5-bromobenzyl bromide 97%
2-Fluoro-5-bromobenzyl bromide

Precaución: For research use only.

Formula: C₇H₅Br₂F
MW: 267.923 g/mol
MDL Number: MFCD09038457
CAS Number: 99725-12-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR