2-Chloro-5-fluoropyridine-1-oxide 97%

Supplier: Ambeed
Warning

A673254-250MG A673254-5G
76831-470EA 40 USD
76831-470 76831-472
2-Chloro-5-fluoropyridine-1-oxide 97%
2-Chloro-5-fluoropyridine-1-oxide

Precaución: for Research Use Only

Formula: C₅H₃ClFNO
MDL Number: MFCD22493425
CAS Number: 405230-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR