2,2,2-Trifluoro-1-(4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazin-1-yl)ethanone 98%

Supplier: Ambeed
Warning

A670111-100MG A670111-1G A670111-250MG
76831-462EA 37.14 USD
76831-462 76831-464 76831-466
2,2,2-Trifluoro-1-(4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazin-1-yl)ethanone 98%
2,2,2-Trifluoro-1-(4-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazin-1-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₈H₂₄BF₃N₂O₃
MW: 384.20 g/mol
MDL Number: MFCD12756469
CAS Number: 1218791-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR