Thiazole-2(3H)-thione 95%
Supplier: Ambeed
|
Danger
|
A668171-1G
A668171-5G
A668171-25G
76831-452EA
43.5
USD
76831-452
77614-062
77684-569
Thiazole-2(3H)-thione 95%
Thiazole-2(3H)-thione
Precaución: For research use only.
Formula:
C₃H₃NS₂ MW: 117.19 g/mol |
MDL Number:
MFCD00022449 CAS Number: 5685-05-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...