Tetraphenylphosphonium phenolate 99+% mix

Supplier: Ambeed
Danger

A666485-10G A666485-25G
76831-448EA 17 USD
76831-448 76831-450
Tetraphenylphosphonium phenolate 99+% mix
Tetraphenylphosphonium phenolate

Precaución: for Research Use Only

Formula: C₃₀H₂₅OP
MW: 432.49 g/mol
MDL Number: MFCD22200717
CAS Number: 15464-47-8
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR