5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)porphyrin 98%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)-21H,23H-porphine, 5,10,15,20-Tetrakis(3,5-dimethoxyphenyl ) -21H,23H-porphyrin

A665060-100MG A665060-1G A665060-250MG
76831-436EA 100.79 USD
76831-436 76831-438 76831-440
5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)porphyrin 98%
5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)porphyrin

Precaución: for Research Use Only

Formula: C₅₂H₄₆N₄O₈
MW: 854.96 g/mol
MDL Number: MFCD02093501
CAS Number: 74684-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR