3-Hydroxy-[1,1'-biphenyl]-4,4'-dicarbonitrile 96%

Supplier: Ambeed
Warning

A665032-100MG A665032-1G A665032-250MG
76831-430EA 50.93 USD
76831-430 76831-432 76831-434
3-Hydroxy-[1,1'-biphenyl]-4,4'-dicarbonitrile 96%
3-Hydroxy-[1,1'-biphenyl]-4,4'-dicarbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₈N₂O
MW: 220.22 g/mol
MDL Number: MFCD18314169
CAS Number: 1261973-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR