4-(Bromomethyl)piperidine hydrochloride 98%

Supplier: Ambeed
Warning

A664639-250MG A664639-1G A664639-5G
76831-428EA 14 USD
76831-428 77614-050 77886-575
4-(Bromomethyl)piperidine hydrochloride 98%
4-(Bromomethyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃BrClN
MDL Number: MFCD09800401
CAS Number: 1159825-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR