4-Carboxy-2-fluorophenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Carboxy-2-fluorophenylboronic Acid Pinacol Ester, 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester
A661249-100MG
A661249-1G
A661249-250MG
A661249-5G
A661249-25G
76831-410EA
8.49
USD
76831-410
76831-412
76831-414
76831-416
77684-557
4-Carboxy-2-fluorophenylboronic acid pinacol ester 98%
4-Carboxy-2-fluorophenylboronic acid pinacol ester
Precaución: For research use only.
Formula:
C₁₃H₁₆BFO₄ MW: 266.07 g/mol |
MDL Number:
MFCD12756466 CAS Number: 1050423-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...