5-Methoxybenzo[d]oxazole 95%

Supplier: Ambeed
Warning

A658643-250MG A658643-5G
76831-402EA 99.73 USD
76831-402 76831-404
5-Methoxybenzo[d]oxazole 95%
5-Methoxybenzo[d]oxazole

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.14 g/mol
MDL Number: MFCD11111707
CAS Number: 132227-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR