(S)-(-)-3-Phenylcyclohexanone 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Phenylcyclohexanone

A658123-100MG A658123-250MG A658123-50MG
76831-396EA 271.6 USD
76831-396 76831-398 76831-400
(S)-(-)-3-Phenylcyclohexanone 98%
(S)-(-)-3-Phenylcyclohexanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄O
MW: 174.2390 g/mol
MDL Number: MFCD08276411
CAS Number: 57344-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR