4-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-((trans-4-Hydroxycyclohexyl)carbamoyl)phenyl)boronic acid

A657306-100MG A657306-1G A657306-250MG A657306-5G
76831-380EA 37.14 USD
76831-380 76831-382 76831-384 76831-386
4-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronic acid 98%
4-(trans-4-Hydroxycyclohexylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MW: 263.0973 g/mol
MDL Number: MFCD09878333
CAS Number: 957062-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR