Methyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate 97%

Supplier: Ambeed
Warning

A656510-1G A656510-250MG A656510-5G
76831-364EA 105.03 USD
76831-364 76831-366 76831-368
Methyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate 97%
Methyl 4'-hydroxy-[1,1'-biphenyl]-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MW: 228.243 g/mol
MDL Number: MFCD01814566
CAS Number: 40501-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR