5-Ethynyl-1,2,3-trifluorobenzene 95%

Supplier: Ambeed
Danger

Synonyms: 3,4,5-Trifluorophenylacetylene

A655299-1G A655299-250MG A655299-5G
76831-358EA 37 USD
76831-358 76831-360 76831-362
5-Ethynyl-1,2,3-trifluorobenzene 95%
5-Ethynyl-1,2,3-trifluorobenzene

Precaución: for Research Use Only

Formula: C₈H₃F₃
MDL Number: MFCD08458177
CAS Number: 158816-55-8
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR