4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: RS-127445

A654799-10MG A654799-1MG A654799-50MG A654799-5MG
76831-344EA 43.5 USD
76831-344 76831-346 76831-348 76831-350
4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine 98%
4-(4-Fluoronaphthalen-1-yl)-6-isopropylpyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆FN₃
MW: 281.32 g/mol
MDL Number: MFCD20270616
CAS Number: 199864-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR