4-Nitro-1,3-dihydro-2H-benzimidazol-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitro-1H-benzo[d]imidazol-2(3H)-one

A654375-100MG A654375-250MG A654375-5G A654375-1G
76831-338EA 14.86 USD
76831-338 76831-340 76831-342 77684-545
4-Nitro-1,3-dihydro-2H-benzimidazol-2-one 98%
4-Nitro-1,3-dihydro-2H-benzimidazol-2-one

Precaución: For research use only.

Formula: C₇H₅N₃O₃
MW: 179.14 g/mol
MDL Number: MFCD09835484
CAS Number: 85330-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR