3-Amino-N-methylpyrazine-2-carboxamide 95%

Supplier: Ambeed
Warning

A654162-1G A654162-250MG
76831-328EA 110.34 USD
76831-328 76831-330
3-Amino-N-methylpyrazine-2-carboxamide 95%
3-Amino-N-methylpyrazine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₈N₄O
MW: 152.15 g/mol
MDL Number: MFCD09953009
CAS Number: 36204-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR