WAY-362450 98+%

Supplier: Ambeed
Warning

Synonyms: WAY 362450, WAY362450, XL-335, Isopropyl 3-(3,4-difluorobenzoyl)-1,1-dimethyl-1,2,3,6-tetrahydroazepino[4,5-b]indole-5-carboxylate, XL335, WAY-362450, propan-2-yl 3-(3,4-difluorobenzoyl)-1,1-dimethyl-2,6-dihydroazepino[4,5-b]indole-5-carboxylate, Turofexorate isopropyl

A653721-100MG A653721-10MG A653721-1MG A653721-25MG A653721-50MG A653721-5MG
76831-310EA 214.31 USD
76831-310 76831-312 76831-314 76831-316 76831-318 76831-320
WAY-362450 98+%
WAY-362450

Precaución: for Research Use Only

Formula: C₂₅H₂₄F₂N₂O₃
MW: 438.47 g/mol
MDL Number: MFCD13181507
CAS Number: 629664-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR