(3-Methoxy-4-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A652949-1G A652949-250MG
76831-302EA 155.96 USD
76831-302 76831-304
(3-Methoxy-4-(trifluoromethyl)phenyl)boronic acid 97%
(3-Methoxy-4-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
MDL Number: MFCD13194426
CAS Number: 1004775-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR