(11bR)-4-Hydroxy-2,6-di(naphthalen-1-yl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥98%, ee 99%

Supplier: Ambeed
Warning

A651569-100MG A651569-1G A651569-250MG
76831-294EA 46 USD
76831-294 76831-296 76831-298
(11bR)-4-Hydroxy-2,6-di(naphthalen-1-yl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥98%, ee 99%
(11bR)-4-Hydroxy-2,6-di(naphthalen-1-yl)-8,9,10,11,12,13,14,15-octahydrodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₄₀H₃₃O₄P
MDL Number: MFCD27952572
CAS Number: 1242066-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR