(R)-2,2'-Bis(bromomethyl)-1,1'-binaphthalene 97%, Ambeed.Inc

Supplier: Ambeed
Danger

A649240-1G A649240-250MG A649240-5G
76831-274EA 186 USD
76831-274 76831-276 77684-534
(R)-2,2'-Bis(bromomethyl)-1,1'-binaphthalene 97%, Ambeed.Inc
(R)-2,2'-Bis(bromomethyl)-1,1'-binaphthalene

Precaución: For research use only.

Formula: C₂₂H₁₆Br₂
MW: 440.17 g/mol
MDL Number: MFCD00185730
CAS Number: 86631-56-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR