1-(2,5-Dichloro-3,6-dimethylphenyl)ethanone 97%

Supplier: Ambeed
Warning

A647337-1G A647337-250MG A647337-25G A647337-5G
76831-250EA 11 USD
76831-250 76831-252 76831-254 76831-256
1-(2,5-Dichloro-3,6-dimethylphenyl)ethanone 97%
1-(2,5-Dichloro-3,6-dimethylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O
MDL Number: MFCD17779313
CAS Number: 164165-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR