(R)-(1,4-Dioxan-2-yl)methanol hydrate 95%

Supplier: Ambeed
Warning

A643656-250MG A643656-1G
76831-246EA 56.23 USD
76831-246 76844-532
(R)-(1,4-Dioxan-2-yl)methanol hydrate 95%
(R)-(1,4-Dioxan-2-yl)methanol hydrate

Precaución: for Research Use Only

Formula: C₅H₁₂O₄
MW: 136.14 g/mol
MDL Number: MFCD31916220
CAS Number: 2829292-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR