2,5-Dichloro-4-nitrophenol 98+%

Supplier: Ambeed
Danger

A638809-1G A638809-250MG A638809-5G
76831-224EA 57.29 USD
76831-224 76831-226 76831-228
2,5-Dichloro-4-nitrophenol 98+%
2,5-Dichloro-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₃
MW: 207.99 g/mol
MDL Number: MFCD08689866
CAS Number: 5847-57-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR