4-Bromo-2-chloro-6-fluoroaniline 95%

Supplier: Ambeed
Warning

A637493-1G A637493-25G A637493-5G
76831-210EA 18.04 USD
76831-210 76831-212 76831-214
4-Bromo-2-chloro-6-fluoroaniline 95%
4-Bromo-2-chloro-6-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MW: 224.46 g/mol
MDL Number: MFCD09878163
CAS Number: 885453-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR