Methyl 4-amino-3-(methylamino)benzoate 98%

Supplier: Ambeed
Warning

A635866-1G A635866-250MG A635866-5G
76831-200EA 108.22 USD
76831-200 76831-202 77684-520
Methyl 4-amino-3-(methylamino)benzoate 98%
Methyl 4-amino-3-(methylamino)benzoate

Precaución: For research use only.

Formula: C₉H₁₂N₂O₂
MW: 180.20 g/mol
MDL Number: MFCD15144544
CAS Number: 616224-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR