4-Chloro-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A633093-10G A633093-1G A633093-25G A633093-5G
76831-186EA 234 USD
76831-186 76831-188 76831-190 76831-192
4-Chloro-N,N-dimethylbenzamide 98%
4-Chloro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MDL Number: MFCD00463859
CAS Number: 14062-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR