2-Bromo-6-methylisonicotinic acid 95%

Supplier: Ambeed
Danger

A632277-100MG A632277-250MG
76831-176EA 43.5 USD
76831-176 76831-178
2-Bromo-6-methylisonicotinic acid 95%
2-Bromo-6-methylisonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.032 g/mol
MDL Number: MFCD13188656
CAS Number: 25462-84-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR