trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronic acid pinacol ester 95% stabilized

Supplier: Ambeed
Warning

Synonyms: (E)-4,4,5,5-Tetramethyl-2-(3-((tetrahydro-2H-pyran-2-yl)oxy)prop-1-en-1-yl)-1,3,2-dioxaborolane

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A630869-1G A630869-250MG A630869-5G
76831-170EA 201.58 USD
76831-170 76831-172 76831-174
trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronic acid pinacol ester 95% stabilized
trans-3-(Tetrahydropyran-2-yloxy)propen-1-ylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₅BO₄
MW: 268.16 g/mol
MDL Number: MFCD03788736
CAS Number: 642066-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR