3-Bromo-2-hydroxy-5-methylbenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A630206-1G
A630206-250MG
A630206-5G
76831-162EA
72
USD
76831-162
76831-164
76831-166
3-Bromo-2-hydroxy-5-methylbenzaldehyde 98%
3-Bromo-2-hydroxy-5-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₇BrO₂ |
MDL Number:
MFCD00275688 CAS Number: 33172-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...