2-Hydroxy-4-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A628270-5G A628270-250MG A628270-1G A628270-25G
76831-150EA 49 USD
76831-150 77613-942 77886-511 77886-512
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid 98%
2-Hydroxy-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₃
MDL Number: MFCD03094996
CAS Number: 1072951-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR