5-Bromo-6-(trifluoromethyl)pyrimidin-4(1H)-one 95%

Supplier: Ambeed
Warning

A627484-1G A627484-5G
76831-146EA 24.41 USD
76831-146 76831-148
5-Bromo-6-(trifluoromethyl)pyrimidin-4(1H)-one 95%
5-Bromo-6-(trifluoromethyl)pyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₅H₂BrF₃N₂O
MW: 242.98 g/mol
MDL Number: MFCD23140968
CAS Number: 126538-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR