2,5-Dimethoxybenzyl 3-methylbutanoate 97%

Supplier: Ambeed
Warning

A624319-10G A624319-5G
76831-136EA 91 USD
76831-136 76831-138
2,5-Dimethoxybenzyl 3-methylbutanoate 97%
2,5-Dimethoxybenzyl 3-methylbutanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₄
MDL Number: MFCD23135509
CAS Number: 876665-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR