N-Methylpyrazin-2-amine 95%

Supplier: Ambeed
Warning

A623976-1G A623976-250MG A623976-5G
76831-130EA 15 USD
76831-130 76831-132 76831-134
N-Methylpyrazin-2-amine 95%
N-Methylpyrazin-2-amine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MDL Number: MFCD08456680
CAS Number: 32111-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR