Decane-2,9-dione 95%

Supplier: Ambeed
Warning

A623961-100MG A623961-250MG
76831-126EA 8 USD
76831-126 76831-128
Decane-2,9-dione 95%
Decane-2,9-dione

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MDL Number: MFCD03413036
CAS Number: 16538-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR