2-(3-Chloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-methylphenylboronic acid pinacol ester

A620959-1G A620959-250MG A620959-5G
76831-118EA 60.48 USD
76831-118 76831-120 76831-122
2-(3-Chloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Chloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₂
MW: 252.55 g/mol
MDL Number: MFCD09972177
CAS Number: 445303-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR