(2,5-Difluoro-1,4-phenylene)diboronic acid 96%

Supplier: Ambeed
Warning

A620689-100MG A620689-1G A620689-250MG A620689-5G
76831-108EA 37.14 USD
76831-108 76831-110 76831-112 76831-114
(2,5-Difluoro-1,4-phenylene)diboronic acid 96%
(2,5-Difluoro-1,4-phenylene)diboronic acid

Precaución: for Research Use Only

Formula: C₆H₆B₂F₂O₄
MW: 201.73 g/mol
MDL Number: MFCD17015780
CAS Number: 1256358-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR