Methyl benzofuran-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A620067-100MG
A620067-1G
A620067-250MG
A620067-5G
76831-098EA
8
USD
76831-098
76831-100
76831-102
76831-104
Methyl benzofuran-6-carboxylate 95%
Methyl benzofuran-6-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₈O₃ |
MDL Number:
MFCD11977827 CAS Number: 588703-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...