6-(4-Fluoro-3-methoxyphenyl)picolinic acid 95%

Supplier: Ambeed
Warning

A619299-100MG A619299-1G A619299-250MG A619299-5G
76831-090EA 25.47 USD
76831-090 76831-092 76831-094 76831-096
6-(4-Fluoro-3-methoxyphenyl)picolinic acid 95%
6-(4-Fluoro-3-methoxyphenyl)picolinic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₃
MW: 247.22 g/mol
MDL Number: MFCD18317543
CAS Number: 1261977-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR