(3-Chloro-4-(chlorocarbonyl)phenyl)boronic acid 95%

Supplier: Ambeed
Danger

A617526-1G A617526-250MG A617526-5G
76831-072EA 145 USD
76831-072 76831-074 76831-076
(3-Chloro-4-(chlorocarbonyl)phenyl)boronic acid 95%
(3-Chloro-4-(chlorocarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BCl₂O₃
MDL Number: MFCD23701457
CAS Number: 850589-38-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR