2-Aminobenzothiazole-6-carboxylic Acid Hydrochloride 98%

Supplier: Ambeed
Warning

A617108-1G A617108-250MG A617108-5G
76831-066EA 20.16 USD
76831-066 76831-068 76831-070
2-Aminobenzothiazole-6-carboxylic Acid Hydrochloride 98%
2-Aminobenzothiazole-6-carboxylic Acid Hydrochloride

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₂S
MW: 230.67 g/mol
MDL Number: MFCD01333078
CAS Number: 18330-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR