Ethylboronic acid pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethane, Ethylboronic acid Pinacol ester

A616511-1G A616511-5G A616511-25G
76831-064EA 6 USD
76831-064 77573-006 77613-914
Ethylboronic acid pinacol ester 95%
Ethylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₈H₁₇BO₂
MDL Number: MFCD19441153
CAS Number: 82954-89-0
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR