2-Bromo-1,3-dimethyl-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A615922-100MG A615922-1G A615922-250MG A615922-5G
76831-056EA 9 USD
76831-056 76831-058 76831-060 76831-062
2-Bromo-1,3-dimethyl-4-nitrobenzene 97%
2-Bromo-1,3-dimethyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MDL Number: MFCD11878621
CAS Number: 60956-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR