BIIB 021 ≥98%

Supplier: Ambeed
Warning

Synonyms: BIIB021, 6-Chloro-9-((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)-9H-purin-2-amine

A615310-10MG A615310-50MG
76831-052EA 45.62 USD
76831-052 76831-054
BIIB 021 ≥98%
BIIB 021

Precaución: for Research Use Only

Formula: C₁₄H₁₅ClN₆O
MW: 318.76 g/mol
MDL Number: MFCD15528939
CAS Number: 848695-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR